Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR41 Monoclonal Antibody (1F3)
GREENER_CHOICE

Catalog No. PIMA522129
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522129 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522129 Supplier Invitrogen™ Supplier No. MA522129
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MLRWLIGGGR EPQGLAEKSP LQTIGEEQTQ NPYTELLVLK AHHDIVRFLV QLDDYRFASA GDDGIVVVWN AQTGEKLLEL NGHTQKITAI ITFPSLESCE EKNQLILTAS ADRTVIVWDG DTTRQVQRIS CFQSTVKCLT VLQRLDVWLS GGNDLCVWNR KLDLLCKTSH LSDTGISALV EIPKNCVVAA VGKELIIFRL VAPTEGSLEW DILEVKRLLD HQDNILSLIN VNDLSFVTGS HVGELIIWDA LDWTMQAYER NFWDPSPQLD TQQEIKLCQK SNDISIHHFT CDEENVFAAV GRGLYVYSLQ MKRVIACQKT AHDSNVLHVA RLPNRQLISC SEDGSVRIWE LREKQQLAAE PVPTGFFNMW GFGRVSKQAS QPVKKQQENA TSCSLELIGD LIGHSSSVEM FLYFEDHGLV TCSADHLIIL WKNGERESGL RSLRLFQKLE ENGDLYLAV.

Non-catalytic component of the C9orf72-SMCR8 complex, a complex that has guanine nucleotide exchange factor (GEF) activity and regulates autophagy. The C9orf72-SMCR8 complex promotes the exchange of GDP to GTP, converting inactive GDP-bound RAB8A and RAB39B into their active GTP-bound form, thereby promoting autophagosome maturation. As part of the C9orf72-SMCR8 complex, stimulates RAB8A and RAB11A GTPase activity in vitro, however WDR42 is shown not be an essential complex component for this function. The C9orf72-SMCR8 complex also acts as a negative regulator of autophagy initiation by interacting with the ULK1/ATG1 kinase complex and inhibiting its protein kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR41
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F3
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene WDR41
Gene Accession No. Q9HAD4
Gene Alias B830029I03Rik; MSTP048; WD repeat domain 41; WD repeat-containing protein 41; WDR41
Gene Symbols WDR41
Host Species Mouse
Immunogen WDR41 (AAH40241, 1 a.a. ~ 459 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55255
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.