Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR42C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535082
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535082 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535082 Supplier Invitrogen™ Supplier No. PA535082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WDR42C Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR42C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DCAF8L2
Gene Accession No. P0C7V8
Gene Alias DCAF8L2; DDB1 and CUL4 associated factor 8 like 2; DDB1 and CUL4 associated factor 8-like 2; DDB1- and CUL4-associated factor 8-like protein 2; WD repeat domain 42C; WD repeat-containing protein 42C; WDR42C
Gene Symbols DCAF8L2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 112-140 amino acids from the N-terminal region of human WDR42C.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347442
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.