Learn More
Invitrogen™ WDR45 Polyclonal Antibody

Description
The antibody detects endogenous levels of total WDR45 protein.
Specifications
Specifications
| Antigen | WDR45 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | WDR45 |
| Gene Accession No. | Q9Y484 |
| Gene Alias | JM5; NBIA4; NBIA5; neurodegeneration with brain iron accumulation 4; neurodegeneration with brain iron accumulation 5; WD repeat domain 45; WD repeat domain phosphoinositide-interacting protein 4; WD repeat domain, X-linked 1; WD repeat-containing protein 45; WD45 repeat protein interacting with phosphoinositides 4; WDR45; WDRX1; WDRXI4; WIPI4; WIPI-4 |
| Gene Symbols | WDR45 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.