Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDR45 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577801
Description
A 1:1,000 dilution of PA5-77801 was sufficient for detection of WIPI4 in 15 μg of Human HeLa Cell lysates by ECL immunoblot analysis using goat anti-rabbit IgG:HRP as the secondary antibody. Detects approximately 40 kDa.
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene has a pseudogene at chromosome 4q31.3. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity and full-length nature of some variants have not been determined.
Specifications
| WDR45 | |
| Polyclonal | |
| Unconjugated | |
| WDR45 | |
| JM5; NBIA4; NBIA5; neurodegeneration with brain iron accumulation 4; neurodegeneration with brain iron accumulation 5; WD repeat domain 45; WD repeat domain phosphoinositide-interacting protein 4; WD repeat domain, X-linked 1; WD repeat-containing protein 45; WD45 repeat protein interacting with phosphoinositides 4; WDR45; WDRX1; WDRXI4; WIPI4; WIPI-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11152, 302559 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
| Q5U2Y0, Q9Y484 | |
| WDR45 | |
| Synthetic peptide from the C-terminal of Human WIPI4 (aa 312-322). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction