Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121118
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121118 Supplier Invitrogen™ Supplier No. PA5121118
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse skeletal muscle, Rat lung. Immunogen sequence: EPLMEKGHLD HEQVGSMGLV EMLHRSNLLA LVGGGSSPKF SEISVLIWDD AREGKDSKEK LVLEFTFTKP VLSVRMRHDK IVIVLKNRIY VYSFPDNPRK LFEFDTRDNP KGLC.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene has a pseudogene at chromosome 4q31.3. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity and full-length nature of some variants have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR45
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene WDR45
Gene Accession No. Q5U2Y0, Q91VM3
Gene Alias JM5; NBIA4; NBIA5; neurodegeneration with brain iron accumulation 4; neurodegeneration with brain iron accumulation 5; WD repeat domain 45; WD repeat domain phosphoinositide-interacting protein 4; WD repeat domain, X-linked 1; WD repeat-containing protein 45; WD45 repeat protein interacting with phosphoinositides 4; WDR45; WDRX1; WDRXI4; WIPI4; WIPI-4
Gene Symbols WDR45
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 35-148 of human WDR45 (NP_001025067.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 302559, 54636
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.