Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDR45B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568658
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
This gene encodes a member of the WIPI or SVP1 family of WD40 repeat-containing proteins. The protein contains seven WD40 repeats that are thought to fold into a beta-propeller structure that mediates protein-protein interactions, and a conserved motif for interaction with phospholipids. The human genome contains several pseudogenes of this gene.
Specifications
WDR45B | |
Polyclonal | |
Unconjugated | |
WDR45B | |
0610008N23Rik; AA408735; D16Bwg0193e; RGD1305141; WD repeat domain 45B; WD repeat domain phosphoinositide-interacting protein 3; WD repeat protein 45-like; WD repeat-containing protein 45B; WD repeat-containing protein 45-like; Wdr45 like; WDR45B; Wdr45l; WDR45-like; WDR45-like protein; WIPI3; WIPI-3; WIPI49-like protein; zgc:56002 | |
Rabbit | |
Affinity chromatography | |
RUO | |
56270 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5MNZ6 | |
WDR45B | |
synthetic peptide directed towards the middle region of human WDR45B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction