Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570601
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570601 Supplier Invitrogen™ Supplier No. PA570601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%.

WDR46 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR46
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene WDR46
Gene Accession No. O15213
Gene Alias 2310007I04Rik; Bing4; C6orf11; C78447; C78559; chromosome 6 open reading frame 11; FP221; putative C6orf11 protein; putative WD repeat domain 46; UTP7; WD repeat domain 46; WD repeat-containing protein 46; WD repeat-containing protein 46; LOW QUALITY PROTEIN: WD repeat-containing protein 46; WD repeat-containing protein BING4; Wdr46
Gene Symbols WDR46
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human WDR46.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9277
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.