Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106388
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106388 Supplier Invitrogen™ Supplier No. PA5106388
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total WDR46.

WDR46 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR46
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene WDR46
Gene Accession No. O15213, Q9Z0H1
Gene Alias 2310007I04Rik; Bing4; C6orf11; C78447; C78559; chromosome 6 open reading frame 11; FP221; putative C6orf11 protein; putative WD repeat domain 46; UTP7; WD repeat domain 46; WD repeat-containing protein 46; WD repeat-containing protein 46; LOW QUALITY PROTEIN: WD repeat-containing protein 46; WD repeat-containing protein BING4; Wdr46
Gene Symbols WDR46
Host Species Rabbit
Immunogen A synthesized peptide derived from human WDR46(Accession O15213), corresponding to amino acid residues F409-E459.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309628, 57315, 9277
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.