Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144857 Supplier Invitrogen™ Supplier No. PA5144857
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSTRTLPHGA GHLAFSQRGL LVAGMGDVVN IWAGQGKASP PSLEQPYLTH RLSGPVHGLQ FCPFEDVLGV GHTGGITSML V Antigen sequence identity: Mouse 94%; Rat 94%.

WDR46 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR46
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR46
Gene Accession No. O15213
Gene Alias 2310007I04Rik; Bing4; C6orf11; C78447; C78559; chromosome 6 open reading frame 11; FP221; putative C6orf11 protein; putative WD repeat domain 46; UTP7; WD repeat domain 46; WD repeat-containing protein 46; WD repeat-containing protein 46; LOW QUALITY PROTEIN: WD repeat-containing protein 46; WD repeat-containing protein BING4; Wdr46
Gene Symbols WDR46
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 395-475. Recombinant protein control fragment (Product #RP-110242).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9277
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.