Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR48 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117073 Supplier Invitrogen™ Supplier No. PA5117073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-251MG, A-431, SGC-7901, LO2, A-549, HT-1080, Mouse brain, Rat testis Immunogen sequence: FSSPDGSDPK LNLGGLLLQA LLEYWPRTHV NPMDEEENEV NHVNGEQENR VQKGNGYFQV PPHTPVIFGE AGGRTLFRLL CRDSGGETES MLLNETVPQW VIDITVDKNM PKFNKIPFYL QPHASSGAKT LKKDRLSASD MLQVRKVMEH VYEKIINLDN ESQTTSSSNN EKPGEQEKEE DIAVLAEEKI ELLCQDQVLD PNMDLRTVKH FIWKSGGDLT LHYRQKST.

WD-repeats are motifs that are found in a variety of proteins and are characterized by a conserved core of 40-60 amino acids, which commonly form a tertiary propeller structure. While proteins that contain WD-repeats participate in a wide range of cellular functions, they are generally involved in regulatory mechanisms involving signal transduction, apoptosis, transcriptional regulation and cell cycle control. WD repeats serve as sites for protein-protein interaction and some seem to mediate the assembly of protein complexes. With eight WD repeats, WDR48 (WD repeat-containing protein 48), also known as USP1-associated factor 1 and p80, is a 677 amino acid protein that functions to regulate deubiquitinating complexes via activation of USP1, USP12 and USP46. WDR48 enhances deubiquitination by increasing catalytic turnover without increasing the affinity of deubiquitinating enzymes for the substrate. WDR48 is ubiquitously expressed and is mainly localized to the cytoplasm. There are five isoforms of WDR48 that are expressed as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR48
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene WDR48
Gene Accession No. Q8BH57, Q8TAF3
Gene Alias 8430408H12Rik; KIAA1449; mKIAA1449; P80; RGD1309702; SPG60; testicular tissue protein Li 224; Uaf1; USP1 associated factor 1; USP1-associated factor 1; WD repeat domain 48; WD repeat endosomal protein; WD repeat-containing protein 48; WDR48
Gene Symbols WDR48
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 450-677 of human WDR48 (NP_0658901).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363164, 57599, 67561
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.