Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDR48 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576203
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
WD-repeats are motifs that are found in a variety of proteins and are characterized by a conserved core of 40-60 amino acids, which commonly form a tertiary propeller structure. While proteins that contain WD-repeats participate in a wide range of cellular functions, they are generally involved in regulatory mechanisms involving signal transduction, apoptosis, transcriptional regulation and cell cycle control. WD repeats serve as sites for protein-protein interaction and some seem to mediate the assembly of protein complexes. With eight WD repeats, WDR48 (WD repeat-containing protein 48), also known as USP1-associated factor 1 and p80, is a 677 amino acid protein that functions to regulate deubiquitinating complexes via activation of USP1, USP12 and USP46. WDR48 enhances deubiquitination by increasing catalytic turnover without increasing the affinity of deubiquitinating enzymes for the substrate. WDR48 is ubiquitously expressed and is mainly localized to the cytoplasm. There are five isoforms of WDR48 that are expressed as a result of alternative splicing events.
Specifications
WDR48 | |
Polyclonal | |
Unconjugated | |
WDR48 | |
8430408H12Rik; KIAA1449; mKIAA1449; P80; RGD1309702; SPG60; testicular tissue protein Li 224; Uaf1; USP1 associated factor 1; USP1-associated factor 1; WD repeat domain 48; WD repeat endosomal protein; WD repeat-containing protein 48; WDR48 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
363164, 57599, 67561 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8BH57, Q8TAF3 | |
WDR48 | |
Recombinant fusion protein of human WDR48. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction