Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR49 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113098
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113098 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113098 Supplier Invitrogen™ Supplier No. PA5113098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WDR49 Polyclonal Antibody for Western Blot, IHC (P), ELISA

WDR49 contains 8 WD repeats. The exact function of WDR49 remains unknown. There are two named isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR49
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene WDR49
Gene Accession No. Q8IV35
Gene Alias WD repeat domain 49; WD repeat-containing protein 49; WDR49
Gene Symbols WDR49
Host Species Rabbit
Immunogen Recombinant Human WD repeat-containing protein 49 protein (251-500AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151790
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.