Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WDR5 0103
GREENER_CHOICE

Catalog No. 532310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-231-0 10 mg
53-235-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-231-0 Supplier Tocris Bioscience™ Supplier No. 5323/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

WDR5 antagonist

WDR5 0103 is a WD repeat-containing protein 5 (WDR5) antagonist (Kd = 450 nM). Disrupts WDR5 interaction with MLL and inhibits MLL core complex methyltransferase activity in vitro. Has no effect on a panel of seven other methyltransferases, including SETD7.

Specifications

Quantity 10 mg
Product Type WDR5 0103
Purity >99%
Molecular Weight (g/mol) 383.44
Inhibitors WDR5 0103

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.