Learn More
Tocris Bioscience™ WDR5 0103

Description
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | WDR5 0103 |
| Purity | >99% |
| Molecular Weight (g/mol) | 383.44 |
| Inhibitors | WDR5 0103 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.