Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR5 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130063AF488
Description
WDR5 Polyclonal antibody specifically detects WDR5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
WDR5 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BIG3, BIG-3, BMP2-induced 3-kb gene protein, SWD3, SWD3, Set1c WD40 repeat protein, homolog, WD repeat domain 5, WD repeat-containing protein 5, WD-repeat protein 5 | |
maps to a region between residue 1 and 50 of human WD repeat domain 5 using the numbering given in entry NP_060058.1 | |
0.1 mL | |
Chromatin Modifiers | |
11091 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction