Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318669
Description
WDR5 Polyclonal antibody specifically detects WDR5 in Human samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ChIP assay (ChIP), ChIP assay SequencingSpecifications
| WDR5 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| BIG3, BIG-3, BMP2-induced 3-kb gene protein, SWD3, SWD3, Set1c WD40 repeat protein, homolog, WD repeat domain 5, WD repeat-containing protein 5, WD-repeat protein 5 | |
| Polyclonal antibody raised in rabbit against human WDR5 (WD (tryptophan-aspartate) repeat domain 5), using a recombinant protein. | |
| 50 μg | |
| Chromatin Modifiers | |
| 11091 | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| ChIP Assay, ChIP sequencing (ChIP-seq), Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/ml | |
| Western Blot 1:1000, Chromatin Immunoprecipitation 2 ug/ChIP, Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:2500 - 1:5000, Chromatin Immunoprecipitation (ChIP), Chromatin Immunoprecipitation Sequencing | |
| P61964 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction