Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR5 Antibody (RAB-C223), Alexa Fluor™ 594 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320133AF594
Description
WDR5 Monoclonal antibody specifically detects WDR5 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
WDR5 | |
Monoclonal | |
Alexa Fluor 594 | |
BIG3, BIG-3, BMP2-induced 3-kb gene protein, SWD3, SWD3, Set1c WD40 repeat protein, homolog, WD repeat domain 5, WD repeat-containing protein 5, WD-repeat protein 5 | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing WDR5 protein under non-denaturing conditions. | |
0.1 mL | |
Chromatin Modifiers | |
11091 | |
Store at 4°C in the dark. | |
IgG κ |
Flow Cytometry, ELISA, Immunofluorescence | |
RAB-C223 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction