Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR5 Mouse anti-Human, Unconjugated, Clone: 2C2, Abnova™

Catalog No. 89335090 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-090 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-090 Supplier Abnova Corporation Supplier No. H00011091M01J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

WDR5 Monoclonal antibody specifically detects WDR5 in Human samples. It is validated for WB-Ce, IF, S-ELISA, ELISA, WB-Re.

Specifications

Antigen WDR5
Applications Immunofluorescence, ELISA, Western Blot
Classification Monoclonal
Clone 2C2
Conjugate Unconjugated
Description WD repeat domain 5
Gene WDR5
Gene Accession No. NM_017588
Gene Alias BIG-3, SWD3
Host Species Mouse
Immunogen WDR5 (NP_060058, 16 a.a. ∼ 120 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 11091
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.