Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR5 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993438
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3438 100 μL
50-199-3439 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3438 Supplier Active Motif Supplier No. 61485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytosolic prostaglandin synthase that catalyzes the oxidoreduction of prostaglandin endoperoxide H2 (PGH2) to prostaglandin E2 (PGE2). Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes. Facilitates HIF alpha proteins hydroxylation via interaction with EGLN1/PHD2, leading to recruit EGLN1/PHD2 to the HSP90 pathway.

Specifications

Antigen WDR5
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene WDR5
Gene Accession No. P61964
Gene Alias 2410008O07Rik; AA408785; AA960360; Big; BIG3; Big-3; BMP2-induced 3-kb gene protein; BOS_12208; CFAP89; cilia and flagella associated protein 89; SWD3; SWD3, Set1c WD40 repeat protein, homolog; WD repeat domain 5; WD repeat-containing protein 5; WD repeat-containing protein BIG-3; Wdr5; wu:fk47f04; zgc:56591; zgc:76895; zgc:76895 protein
Gene Symbols WDR5
Host Species Rabbit
Immunogen This antibody was raised against a peptide within the C-terminal region of human WDR5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11091
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.