Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR5 Rabbit anti-Bovine, Canine, Chicken, Human, Mouse, Non-human Primate, Plant, Porcine, Rat, Xenopus laevis, Zebrafish, Polyclonal, FabGennix

Catalog No. 502209638
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9638 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9638 Supplier Fabgennix Inc Supplier No. WDR5101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WDR5 Polyclonal antibody specifically detects WDR5 in Bovine, Canine, Chicken, Human, Mouse, Non-human Primate, Plant, Porcine, Rat, Xenopus laevis, Zebrafish samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen WDR5
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene WDR5
Gene Accession No. P61964, P61965, Q2KIG2, Q498M4
Gene Alias 2410008O07Rik; AA408785; AA960360; Big; BIG3; BIG-3; BMP2-induced 3-kb gene protein; CFAP89; cilia and flagella associated protein 89; SWD3; SWD3, Set1c WD40 repeat protein, homolog; Unknown (protein for MGC:137327); WD repeat domain 5; WD repeat-containing protein 5; WD repeat-containing protein BIG-3; Wdr5; wu:fk47f04; zgc:56591; zgc:76895; zgc:76895 protein
Host Species Rabbit
Immunogen Synthetic peptide:DVKTGKCLKTLPAHSDPVcorresponding to amino acids 157-174 of Human WDR5 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100125836, 100156729, 11091, 140858, 362093, 406372, 417144, 608113, 746901
Target Species Bovine, Canine, Chicken, Human, Mouse, Non-human Primate, Plant, Pig, Rat, X. laevis, Zebrafish
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.