Learn More
WDR5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 155441AP150UL

Description
The WDR5 antibody targets the WD repeat protein domain 5 (WDR5), a member of the WD40 repeat protein family in humans.
Cytosolic prostaglandin synthase that catalyzes the oxidoreduction of prostaglandin endoperoxide H2 (PGH2) to prostaglandin E2 (PGE2). Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes. Facilitates HIF alpha proteins hydroxylation via interaction with EGLN1/PHD2, leading to recruit EGLN1/PHD2 to the HSP90 pathway.Specifications
WDR5 | |
Polyclonal | |
Unconjugated | |
WDR5 | |
BIG 3, BIG3, BMP2 induced 3 kb gene protein, SWD3, WD repeat containing protein 5, WD repeat domain 5, WDR5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11091, 140858, 362093 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61964, P61965, Q498M4 | |
Wdr5 | |
WDR5 Fusion Protein Ag7900 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.