Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR5, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89998389 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-389 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-389 Supplier Abnova Corporation Supplier No. PAB31340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen WDR5
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide, 0.05% proclin 300).
Gene WD repeat domain 5
Gene Alias BIG-3/SWD3
Gene Symbols WDR5
Host Species Rabbit
Immunogen Recombinant protein corresponding to human WDR5.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11091
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.