Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR5 Recombinant Rabbit Monoclonal Antibody (18D11)
GREENER_CHOICE

Catalog No. FEMA550207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550207 Supplier Invitrogen™ Supplier No. MA550207
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

WDR5 Recombinant Monoclonal Antibody for IHC (P), Flow, ELISA

Cytosolic prostaglandin synthase that catalyzes the oxidoreduction of prostaglandin endoperoxide H2 (PGH2) to prostaglandin E2 (PGE2). Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes. Facilitates HIF alpha proteins hydroxylation via interaction with EGLN1/PHD2, leading to recruit EGLN1/PHD2 to the HSP90 pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR5
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 18D11
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene WDR5
Gene Accession No. P61964
Gene Alias 2410008O07Rik; AA408785; AA960360; Big; BIG3; BIG-3; BMP2-induced 3-kb gene protein; CFAP89; cilia and flagella associated protein 89; SWD3; SWD3, Set1c WD40 repeat protein, homolog; Unknown (protein for MGC:137327); WD repeat domain 5; WD repeat-containing protein 5; WD repeat-containing protein BIG-3; Wdr5; wu:fk47f04; zgc:56591; zgc:76895; zgc:76895 protein
Gene Symbols WDR5
Host Species Rabbit
Immunogen A synthesized peptide derived from Human WDR5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11091
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.