Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR52 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564321 Supplier Invitrogen™ Supplier No. PA564321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSFGYDCRKR ANLQLLDDSI AIYIAGNQLI FLNLKTKEQI YLRSSSGEGI GVIGVHPHKT YFTVAEKGSF PDIIIYEYPS LRPYRVLRDG TEKG Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 73%.

Flagellar protein involved in sperm flagellum axoneme organization and function. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR52
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP44
Gene Accession No. Q96MT7
Gene Alias CFAP44; cilia and flagella associated protein 44; cilia- and flagella-associated protein 44; WD repeat domain 52; WD repeat-containing protein 52; WDR52
Gene Symbols CFAP44
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR52. Recombinant protein control fragment (Product #RP-103654).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55779
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.