Learn More
Invitrogen™ WDR54 Polyclonal Antibody

Description
WDR54 Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | WDR54 |
| Applications | ELISA, Western Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300, 0.02% sodium azide; pH 7.4 |
| Gene | WDR54 |
| Gene Accession No. | Q9H977, Q9R0D8 |
| Gene Alias | 1700030E05Rik; D3Mm3e; WD repeat domain 54; WD repeat-containing protein 54; WDR54 |
| Gene Symbols | WDR54 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.