Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR55 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561665 Supplier Invitrogen™ Supplier No. PA561665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSKAHGAPIN SLLLVDENVL ATGDDTGGIR LWDQRKEGPL MDMRQHEEYI ADMALDPAKK LLLTASGDGC LGIFNIKRRR FELLSEPQSG DLT Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.

WD repeat domain 55 (WDR55) also named as, FLJ20195, is a 383 amino acid protein, which contains seven WD repeats and belongs to the WD repeat WDR55 family. WDR55 localizes in the nucleus and cytoplasm. WDR55 acts as a modulator of rRNA synthesis and play a central role in organogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR55
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR55
Gene Accession No. Q9H6Y2
Gene Alias WD repeat domain 55; WD repeat-containing protein 55; WDR55
Gene Symbols WDR55
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR55. Recombinant protein control fragment (Product #RP-100128).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54853
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.