Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR57 Antibody, Novus Biologicals™
SDP

Catalog No. NB429469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB429469 25 μL
NBP192585 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB429469 Supplier Novus Biologicals Supplier No. NBP19258525UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WDR57 Polyclonal specifically detects WDR57 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen WDR57
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias 38 kDa-splicing factor, HPRP8BP, Prp8-binding protein, PRP8BP40K, PRPF8BPFLJ41108, RP11-490K7.3, SFP38, small nuclear ribonucleoprotein 40kDa (U5), SPF38MGC1910, U5 small nuclear ribonucleoprotein 40 kDa protein, U5 snRNP 40 kDa protein, U5 snRNP-specific 40 kDa protein, U5 snRNP-specific 40 kDa protein (hPrp8-binding), U5-40K, U5-40kD protein, WD repeat domain 57 (U5 snRNP specific), WD repeat-containing protein 57, WDR57
Gene Symbols SNRNP40
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:QGNVHNFEKNLLRCSWSPDGSKIAAGSADRFVYVWDTTSRRILYKLPGHAGSINEVAFHPDEPIIISASSDKRLYMGEIQ
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9410
Test Specificity Specificity of human WDR57 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.