Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDR5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560191
Description
Immunogen sequence: MATKESRDAK AQLALSSSAN QSKEVPE Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 48%.
This intronless gene encodes a protein containing several WD40 repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, including a trp-asp at the C-terminal end. The encoded protein may mediate protein-protein interactions.
Specifications
WDR5B | |
Polyclonal | |
Unconjugated | |
WDR5B | |
WD repeat domain 5B; WD repeat-containing protein 5B; WDR5B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54554 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86VZ2 | |
WDR5B | |
Recombinant protein corresponding to Human WDR5B. Recombinant protein control fragment (Product #RP-99269). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction