Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDR60 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554244
Description
Immunogen sequence: DIQTEEIETR EVWTQHPGES TVVSGGSEQR DTSDAVVMPK IDTPRLCSFL RAACQVMAVL LEEDRLAAEP SWNLRAQDRA LYFSDSSSQL NTSLPFLQNR KVSSLHTSRV QRQMVVSVH Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.
Specifications
| WDR60 | |
| Polyclonal | |
| Unconjugated | |
| WDR60 | |
| FAP163; SRPS6; SRTD8; testicular secretory protein Li 66; WD repeat domain 60; WD repeat-containing protein 60; WDR60 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55112 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WVS4 | |
| WDR60 | |
| Recombinant protein corresponding to Human WDR60. Recombinant protein control fragment (Product #RP-92699). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction