Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR61 Monoclonal Antibody (OTI4D2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA807592

Description
SKI8 is a component of the PAF and SKI complex which is composed of SKI2, SKI3 and SKI8. These are involved in transcriptional regulation and events downstream of RNA synthesis. Interaction of the SKI complex with SKI7 links the SKI complex to the exosome in order to perform mRNA degradation.Specifications
WDR61 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9GZS3 | |
WDR61 | |
Human recombit protein fragment corresponding to amino acids 92-305 of human WDR61 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI4D2 | |
Unconjugated | |
WDR61 | |
REC14, SKI8 | |
Mouse | |
Affinity Chromatography | |
RUO | |
80349 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction