Learn More
Invitrogen™ WDR61 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530833
Description
Recommended positive controls: HepG2, mouse stomach. Predicted reactivity: Mouse (98%), Rat (97%), Zebrafish (90%), Xenopus laevis (89%), Chicken (94%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SKI8 is a component of the PAF and SKI complex which is composed of SKI2, SKI3 and SKI8. These are involved in transcriptional regulation and events downstream of RNA synthesis. Interaction of the SKI complex with SKI7 links the SKI complex to the exosome in order to perform mRNA degradation.
Specifications
WDR61 | |
Polyclonal | |
Unconjugated | |
WDR61 | |
2700038L12Rik; 2810418I05Rik; meiotic recombination protein REC14; meiotic recombination REC14 protein homolog; REC14; recombination protein REC14; RGD1308228; SKI8; SKI8 homolog; WD repeat domain 61; WD repeat-containing protein 61; WD repeat-containing protein 61, N-terminally processed; WDR61 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
66317, 80349 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.6 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9ERF3, Q9GZS3 | |
WDR61 | |
Recombinant protein encompassing a sequence within the center region of human WDR61. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.