Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR61 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559052
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559052 Supplier Invitrogen™ Supplier No. PA559052
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DISHTLPIAA SSSLDAHIRL WDLENGKQIK SIDAGPVDAW TLAFSPDSQY LATGTHVGKV NIFGVESGKK EYSLDTR Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

SKI8 is a component of the PAF and SKI complex which is composed of SKI2, SKI3 and SKI8. These are involved in transcriptional regulation and events downstream of RNA synthesis. Interaction of the SKI complex with SKI7 links the SKI complex to the exosome in order to perform mRNA degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR61
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR61
Gene Accession No. Q4V7A0, Q9ERF3, Q9GZS3
Gene Alias 2700038L12Rik; 2810418I05Rik; meiotic recombination protein REC14; meiotic recombination REC14 protein homolog; REC14; recombination protein REC14; RGD1308228; SKI8; SKI8 homolog; WD repeat domain 61; WD repeat-containing protein 61; WD repeat-containing protein 61, N-terminally processed; WDR61
Gene Symbols WDR61
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR61. Recombinant protein control fragment (Product #RP-98085).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363064, 66317, 80349
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.