Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR61 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572827
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572827 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572827 Supplier Invitrogen™ Supplier No. PA572827
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WDR61 Polyclonal Antibody for Western Blot, IHC (P), ELISA

SKI8 is a component of the PAF and SKI complex which is composed of SKI2, SKI3 and SKI8. These are involved in transcriptional regulation and events downstream of RNA synthesis. Interaction of the SKI complex with SKI7 links the SKI complex to the exosome in order to perform mRNA degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR61
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene WDR61
Gene Accession No. Q4V7A0, Q9ERF3, Q9GZS3
Gene Alias 2700038L12Rik; 2810418I05Rik; meiotic recombination protein REC14; meiotic recombination REC14 protein homolog; REC14; recombination protein REC14; RGD1308228; SKI8; SKI8 homolog; WD repeat domain 61; WD repeat-containing protein 61; WD repeat-containing protein 61, N-terminally processed; WDR61
Gene Symbols WDR61
Host Species Rabbit
Immunogen WDR61 antibody for a 14 amino acid peptide near the amino terminus of human WDR61. The immunogen is within the first 50 amino acids of WDR61.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363064, 66317, 80349
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.