Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR62 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10077302H
Description
WDR62 Polyclonal antibody specifically detects WDR62 in Human, Mouse samples. It is validated for Western BlotSpecifications
WDR62 | |
Polyclonal | |
Western Blot | |
C19orf14MGC166976, chromosome 19 open reading frame 14, DKFZP434J046, DKFZp686G1024, EC 2.7.11.1, EC 2.7.7.38, FLJ33298, WD repeat domain 62, WD repeat-containing protein 62 | |
The immunogen recognized by this antibody maps to a region between residue 750 and 800 of human WD repeat domain 62 using the numbering given in entry NP_775907.4 (GeneID 284403). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
284403 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction