Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR63 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107047
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107047 Supplier Invitrogen™ Supplier No. PA5107047
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%.

WDR63 contains 5 WD repeats. The exact function of WDR63 is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR63
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene WDR63
Gene Accession No. Q8IWG1
Gene Alias DIC3; NYD-SP29; testicular tissue protein Li 225; Testis development protein NYD-SP29; testis development protein NYD-SP29 (NYD-SP29); WD repeat domain 63; WD repeat-containing protein 63; WDR63
Gene Symbols WDR63
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human WDR63 (aa 288-338).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126820
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.