Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR65 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555922
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555922 Supplier Invitrogen™ Supplier No. PA555922
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKLTKKVRPQ EVSETEPSRD MLSTAPTARL NEQEETGRII EMQRLEIQRL RDQIQEQEQV TGFHTLAGVR LPSLSNSEVD LEVKTN Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 28%.

The WDR65 gene, which stands for WD Repeat Domain 65, encodes a protein characterized by the presence of WD repeat domains that facilitate protein-protein interactions. This gene is implicated in the assembly of a subset of inner arm dyneins, critical for ciliary motility and axonemal structure. Mutations in WDR65 have been associated with diseases such as Van der Woude syndrome (VWS) and related craniofacial disorders, highlighting its role in developmental processes. Specifically, a missense mutation in WDR65 was identified as a potential cause of VWS, suggesting that WDR65 is a target for regulatory factors like IRF6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR65
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP57
Gene Accession No. Q96MR6
Gene Alias CFAP57; cilia and flagella associated protein 57; cilia- and flagella-associated protein 57; VWS2; WD repeat-containing protein 65; WDR65
Gene Symbols CFAP57
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR65. Recombinant protein control fragment (Product #RP-94575).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149465
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.