Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR65 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531657 Supplier Invitrogen™ Supplier No. PA531657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat. Predicted reactivity: Mouse (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The WDR65 gene, which stands for WD Repeat Domain 65, encodes a protein characterized by the presence of WD repeat domains that facilitate protein-protein interactions. This gene is implicated in the assembly of a subset of inner arm dyneins, critical for ciliary motility and axonemal structure. Mutations in WDR65 have been associated with diseases such as Van der Woude syndrome (VWS) and related craniofacial disorders, highlighting its role in developmental processes. Specifically, a missense mutation in WDR65 was identified as a potential cause of VWS, suggesting that WDR65 is a target for regulatory factors like IRF6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR65
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CFAP57
Gene Accession No. Q96MR6
Gene Alias CFAP57; cilia and flagella associated protein 57; cilia- and flagella-associated protein 57; VWS2; WD repeat-containing protein 65; WDR65
Gene Symbols CFAP57
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human WDR65. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149465
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.