Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR66 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543884
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543884 Supplier Invitrogen™ Supplier No. PA543884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GELEEKTDRM PQDELGQERR DLEPENREEG QERRVSDIQS KAGISRESLV Sequence homology: Dog: 100%; Human: 100%; Pig: 85%.

WDR66 contains 9 WD repeats. This protein encoded by WDR66 belongs to the WD repeat-containing family of proteins, which function in the formation of protein-protein complexes in a variety of biological pathways. This family member appears to function in the determination of mean platelet volume (MPV), and polymorphisms in this gene have been associated with variance in MPV. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR66
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene WDR66
Gene Accession No. Q8TBY9
Gene Alias CaM-IP4; CFAP251; cilia and flagella associated protein 251; cilia- and flagella-associated protein 251; si:dkey-65b12.4; WD repeat domain 66; WD repeat-containing protein 66; WD repeat-containing protein 66; cilia- and flagella-associated protein 251; WDR66
Gene Symbols WDR66
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human WDR66 (aa 180-229).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144406
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.