Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR68 Antibody, Novus Biologicals™
SDP

Catalog No. NB405324 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB405324 25 μL
NBP192589 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB405324 Supplier Novus Biologicals Supplier No. NBP19258925UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

WDR68 Polyclonal specifically detects WDR68 in Human, Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen WDR68
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P61962
Gene Alias DDB1 and CUL4 associated factor 7, DDB1- and CUL4-associated factor 7, HAN11AN11, human anthocyanin, seven-WD-repeat protein of the AN11 family-1, SWAN-1, WD repeat domain 68, WD repeat-containing protein 68, WD repeat-containing protein An11 homolog, WDR68, WD-repeat protein
Gene Symbols DCAF7
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:PCTPVARLNNHRACVNGIAWAPHSSCHICTAADDHQALIWDIQQMPRAIEDPILAYTAEGEINNVQWASTQPDWIAICYNNC
Molecular Weight of Antigen 39 kDa
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10238
Test Specificity Specificity of human WDR68 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Drosophila
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.