Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR69 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531812 Supplier Invitrogen™ Supplier No. PA531812
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1. Predicted reactivity: Rat (89%), Zebrafish (84%), Xenopus laevis (87%), Chimpanzee (100%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The WDR69 gene, also known as WD Repeat Domain 69, encodes a protein characterized by WD repeat domains. These domains are known for facilitating protein-protein interactions and are involved in various cellular processes. However, specific information regarding the function and clinical relevance of WDR69 remains limited compared to other genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR69
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene DAW1
Gene Accession No. Q8N136
Gene Alias DAW1; dynein assembly factor with WD repeats 1; dynein assembly factor with WDR repeat domains 1; ODA16; outer row dynein assembly 16 homolog; outer row dynein assembly protein 16 homolog; testis tissue sperm-binding protein Li 93mP; WD repeat domain 69; WD repeat-containing protein 69; WDR69
Gene Symbols DAW1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human WDR69. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164781
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.