Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
| Antigen | WDR7 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
WDR7 Polyclonal specifically detects WDR7 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| WDR7 | |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 23335 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: AKVQEEEHDLVDDDITTGCLSSVPQMKKISTSYEERRKQATAIVLLGVIGAEFGAEIEPPKLLTRPRSSSQIPEGFGLTSGGSNYSLARHTCKALTFLLLQPPSPKLPPHS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human | |
| TRAG, WDR7 WD repeat domain 7 | |
| WDR7 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title