Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559854 Supplier Invitrogen™ Supplier No. PA559854
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKVQEEEHDL VDDDITTGCL SSVPQMKKIS TSYEERRKQA TAIVLLGVIG AEFGAEIEPP KLLTRPRSSS QIPEGFGLTS GGSNYSLARH TCKALTFLLL QPPSPKLPPH S Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD) that may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. The encoded protein forms the beta subunit of rabconnectin-3 and binds directly with Rab3A GDP/GTP exchange protein and indirectly with Rab3A GDP/GTP activating protein; these proteins are regulators of Rab3 small G protein family members involved in control of the calcium-dependent exocytosis of neurotransmitters. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR7
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR7
Gene Accession No. Q9Y4E6
Gene Alias AI462727; Kiaa0541; LOW QUALITY PROTEIN: WD repeat-containing protein 7; mKIAA0541; rabconnectin-3 beta; rabconnectin-3beta; TGF-beta resistance associated; TGF-beta resistance-associated protein TRAG; TRAG; WD repeat domain 7; WD repeat-containing protein 7; WD repeat-containing protein 7-like protein; Wd7; WDR7
Gene Symbols WDR7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR7. Recombinant protein control fragment (Product #RP-98239).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23335
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.