Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDR70 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116159
Description
WDR70 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
WDR70 gene ontology annotations related to this gene include regulation of DNA double-strand break processing; regulation of histone H2B conserved C-terminal lysine ubiquitination.
Specifications
| WDR70 | |
| Polyclonal | |
| Unconjugated | |
| WDR70 | |
| FLJ10233; WD repeat domain 70; WD repeat-containing protein 70; WDR70 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 55100 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.45 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9NW82 | |
| WDR70 | |
| KLH conjugated synthetic peptide between 575-604 amino acids from the C-terminal region of human WDR70. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction