Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563438 Supplier Invitrogen™ Supplier No. PA563438
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GHSYIYQLLN SGLSKSIYPA DGRVLKETIY PHLLCSTSVQ ENKEQSRPFV MGYMNERKEP FYKVLFSGEV SGRITLWHIP DVPVSKFDGS PREIPVTATW TLQDNFDKHD TMSQSIIDYF Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

Plays a major role in formation of tooth enamel (PubMed:19853237, PubMed:25008349). Specifically required during the maturation phase of amelogenesis for normal formation of the enamel matrix and clearance of enamel proteins. May be involved in localization of the calcium transporter SLC24A4 to the ameloblast cell membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR72
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR72
Gene Accession No. Q3MJ13
Gene Alias AI2A3; WD repeat domain 72; WD repeat-containing protein 72; WDR72
Gene Symbols WDR72
Host Species Rabbit
Immunogen Recombinant protein fragment of Human WDR72 (Product #RP-98158)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256764
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.