Learn More
Description
Specifications
Specifications
| Antigen | WDR73 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | NP_116245 |
| Gene Alias | FLJ00296 protein, FLJ14888, HSPC264, WD repeat domain 73, WD repeat-containing protein 73 |
| Gene Symbols | WDR73 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
