Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR74 Monoclonal Antibody (OTI4B2)
GREENER_CHOICE

Catalog No. PIMA526448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526448 Supplier Invitrogen™ Supplier No. MA526448
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WDR74 Monoclonal Antibody for Western Blot, IHC (P)

The yeast nucleolar protein NOP7 is necessary for the maturation of 66S preribosomes and interacts with numerous other proteins. One such protein is an essential, conserved WD repeat protein, NOP seven-associated protein 1 (NSA1), that is also required for the yeast 66S ribosome assembly. NSA1 is also associated with the AAA ATPase Rix7, and release of NSA1 from a novel late nucleolar pre-60S requires the Rix7 function. NSA1 has also been found upregulated in mammalian cancer cells, suggesting it may also play a role in cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR74
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene WDR74
Gene Accession No. Q6RFH5, Q8VCG3
Gene Alias 5730436H21Rik; AA407588; NOP seven-associated protein 1; NSA1; WD repeat domain 74; WD repeat-containing protein 74; Wdr74
Gene Symbols WDR74
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 114-385 of WDR74 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107071, 483778, 54663, 690229
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.