Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDR74 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583417
Description
Immunogen sequence: RNDWLDLRVP IWDQDIQFLP GSQKLVTCTG YHQVRVYDPA SPQRRPVLET TYGEYPLTAM TLTPGGNSVI VGNTHGQLAE ID.
The yeast nucleolar protein NOP7 is necessary for the maturation of 66S preribosomes and interacts with numerous other proteins. One such protein is an essential, conserved WD repeat protein, NOP seven-associated protein 1 (NSA1), that is also required for the yeast 66S ribosome assembly. NSA1 is also associated with the AAA ATPase Rix7, and release of NSA1 from a novel late nucleolar pre-60S requires the Rix7 function. NSA1 has also been found upregulated in mammalian cancer cells, suggesting it may also play a role in cell proliferation.
Specifications
WDR74 | |
Polyclonal | |
Unconjugated | |
WDR74 | |
5730436H21Rik; AA407588; NOP seven-associated protein 1; NSA1; WD repeat domain 74; WD repeat-containing protein 74; Wdr74 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54663 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6RFH5 | |
WDR74 | |
Recombinant protein corresponding to Human WDR74. Recombinant protein control fragment (Product #RP-102831). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction