Learn More
WDR77/MEP50 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | WDR77/MEP50 |
---|---|
Applications | Immunofluorescence, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1922
![]() |
Bethyl Laboratories, Inc
IHC00338T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1921
![]() |
Bethyl Laboratories, Inc
IHC00338 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Chimpanzee and Northern white-cheeked gibbon
WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles (Friesen et al., 2002).Specifications
WDR77/MEP50 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
MEP-50; OTTHUMP00000013473; WD repeat-containing protein 77; androgen receptor cofactor p44; methylosome protein 50; p44/Mep50 | |
Between 292 and 342 | |
Primary | |
4° C | |
WDR77 |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q99J09, Q9BQA1 | |
70465, 79084 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.