Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDR77 Monoclonal Antibody (2G9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA532970
Description
Purity is > 95% by SDS-PAGE.
WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles.
Specifications
WDR77 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q4QR85, Q99J09, Q9BQA1 | |
WDR77 | |
Recombinant full length Human MEP50. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry, Western Blot | |
2G9 | |
Unconjugated | |
WDR77 | |
2610003I18Rik; 2610312E17Rik; Ac2-269; androgen receptor cofactor p44; C79984; HKMT1069; MEP50; MEP-50; Methylosome protein 50; MGC2722; Nbla10071; OTTHUMP00000013473; p44; p44/Mep50; RGD1310479; RP11-552M11.3; testis tissue sperm-binding protein Li 44a; WD repeat domain 77; WD repeat-containing protein 77; WD45; WDR77 | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
310769, 70465, 79084 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction