Learn More
WDR77 Monoclonal Antibody (OTI5H9)

Description
WDR77 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | WDR77 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI5H9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | WDR77 |
| Gene Accession No. | Q99J09, Q9BQA1 |
| Gene Alias | 2610003I18Rik; 2610312E17Rik; Ac2-269; androgen receptor cofactor p44; C79984; HKMT1069; MEP50; MEP-50; Methylosome protein 50; MGC2722; Nbla10071; OTTHUMP00000013473; p44; p44/Mep50; RGD1310479; RP11-552M11.3; testis tissue sperm-binding protein Li 44a; WD repeat domain 77; WD repeat-containing protein 77; WD45; WDR77 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.